Learn More
Medchemexpress LLC Enzalutamide, 915087-33-1, MFCD14155804, 50 mg
Supplier: Medchemexpress LLC HY7000250MG
Molecular Formula: C21 H16 F4 N4 O2 S, Molecular Weight: 464.441, 99.71%, Synonyms: MDV3100. An androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.