Learn More
Crescent Chemical Co Inc EPA 200.8 QC STD 20COMP 100ML
SDP
Supplier:  Crescent Chemical Co Inc CTCC5608R.1
Encompass_Preferred
Calibration Standards for Method 200.8 QC Standard 20 20 Components in 5% HNO3 tr. Tartaric acid Ag 10 UgmL Al 10 UgmL As 10 UgmL Ba 10 UgmL Be 10 UgmL Cd 10 UgmL Co 10 ugmL Cr 10 UgmL Cu 10 UgmL Mn 10 UgmL Mo 10 UgmL Ni 10 UgmL Pb 10 UgmL Sb 10 Ugm
Catalog No. 50-685-64
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.