Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epinephrine, USP, 97-100.5%, Spectrum™ Chemical
SDP

Catalog No. p-7157402 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Amber Glass Bottle

EP130, 51-43-4, C9H13NO3

Spectrum™ Chemical Epinephrine, USP is also known as adrenaline and is a hormone and neurotransmitter. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 51-43-4
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.21
InChI Key UCTWMZQNUQWSLP-VIFPVBQESA-N
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNC[C@H](O)C1=CC=C(O)C(O)=C1

Specifications

CAS Max % 100%
Ignition Residue Negligible
Loss on Drying 2%
Packaging Amber Glass Bottle
Quantity 100 g
UN Number UN2811
Specific Rotation -50.0° to -54.0°
Percent Purity 97 to 100.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.