Learn More
AdipoGen Epoxidized soya bean oil
SDP
Supplier:  AdipoGen CDXE0037G050
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 8013-07-8. Synthetic. Useful for the determination of ESBO in foods with HPLC-MS or GC-MS. ESBO is currently employed as plasticizer and HCl scavenger in PVC products. Vegetable oils are widely used for chemical manipulation because of the high numbers of carbon-carbon double bonds. Through the epoxidation, an epoxide group which is a more reactive group than a double bond is added to the soybean oil, transforming it in a good hydrochloric acid scavenger and good plasticizer.
Catalog No. 50-256-0990
missing translation for 'QuantityLeft'