Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER 50891, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Antagonist of RARα receptors (IC50 = 31.2 nM). Displays selective affinity for RARα (relative IC50 values are 1.8, 432 and 535 nM for RARα, RARγ and RARβ respectively).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 432.51 |
Percent Purity | >99% |
Chemical Name or Material | ER 50891 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction