Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERB 041, Tocris Bioscience™

Catalog No. p-4816695 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent ERβ agonist

Potent ERβ agonist; displays >200-fold selectivity for ERβ over ERα (IC50 values are 5 and 1216 nM for human ERβ and ERα; 3.1 and 620 nM for rat ERβ and ERα; and 3.7 and 750 nM for mouse ERβ and ERα respectively).

Chemical Identifiers

CAS 524684-52-4
Molecular Formula C15H10FNO3
Molecular Weight (g/mol) 271.247
InChI Key FCXYSEXZEGPLGG-DHDCSXOGSA-N
Synonym prinaberel, unii-a9c8mnf7ca, a9c8mnf7ca, 2-3-fluoro-4-hydroxyphenyl-7-vinyl-1,3-benzoxazol-5-ol, prinaberel usan:inn, 4-2z-7-ethenyl-5-hydroxy-3h-1,3-benzoxazol-2-ylidene-2-fluorocyclohexa-2,5-dien-1-one, prinaberel usan/inn, bidd:er0139, erb hplc, 7-ethenyl-2-3-fluoro-4-hydroxyphenyl benzoxazol-5-ol
PubChem CID 5326893
IUPAC Name (4Z)-4-(7-ethenyl-5-hydroxy-3H-1,3-benzoxazol-2-ylidene)-2-fluorocyclohexa-2,5-dien-1-one
SMILES C=CC1=CC(=CC2=C1OC(=C3C=CC(=O)C(=C3)F)N2)O

Specifications

Quantity 10 mg
Formula Weight 271.24
Percent Purity >99%
Chemical Name or Material ERB 041
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.