Learn More
Bevco Precision Manufacturing Co ErgoLux Backless Stool, Soft Black Polyurethane Seat, black nylon Base, Dual-Wheel Hard Floor Casters, 18"-22.5" Seat Height Adj.
Supplier:  Bevco Precision Manufacturing Co S3000BLK
Encompass_Preferred
BEVCO's ErgoLux Series advances the backless stool experience with the ultimate in comfort and durability. It is ideal for a wide range of applications, including general office, medical, laboratory, and higher education applications.The stress-relieving qualities of the stool are heightened because they respond to your body’s unique shape, weight, and temperature.The 3.5" thick ultrasoft, high-density, visco-elastic memory foam seat provides resilient, long-lasting ergonomic support.The 14.5" diameter seat is covered by puncture, water, and stain-resistant polyurethane that is non-slip and easy to clean.Easy-to-use pneumatic height adjustment allows you to change your seat height from 18" - 22.5".This stool comes with a sturdy 25" diameter black nylon base.Model comes standard with dual-wheel hard floor casters.
Catalog No. NC2119023
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.