Learn More
World Precision Instrument Ergonomic Micro-Shear Flush Cutters, 12.7 cm, Polished Blades
SDP
Supplier:  World Precision Instrument 504751
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Size and shape of these cutters are optimized to work in high density areas. Cutting capacity: 16 AWG (1.29 mm) soft wire.
Catalog No. 50-253-9293
missing translation for 'QuantityLeft'