Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foamtec ErgoWRENCH™ Vacuum Chamber Scrubbing Tools: Heads

Catalog No. p-4370276 Shop All Foamtec International Products
Click to view available options
Product Type:
Size:
Format:

For safer and more efficient scrubbing of hard to reach areas throughout vacuum chambers

Stiff swivel head can be adjusted to optimize scrubbing angles. Different size heads are interchangeable, which allows the ErgoWRENCH to be adjusted for different applications.

  • Reduces scrub-time by focusing Diamond ScrubPAD directly onto heavy deposition buildup, quickly taking chamber back to bare metal
  • Removes heavy deposition from areas critical to production, preventing unwanted flaking
  • Safely removes deposition from sensitive areas
  • Quick removal of deposition from aluminum, stainless-steel, ceramic, glass, quartz and anodized surfaces while minimizing tool wear

Specifications

Product Type

Scrub Head

Format

Non-removable, Fixed

For Use With (Application)

For safer and more efficient scrubbing of hard to reach areas

For Use With (Equipment)

Interchangeable handles

Includes

Hook

Material

Delrin™ Plastic

Size

Medium

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.