Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erio Green B, TCI America™
SDP

Catalog No. A06875G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 12768-78-4
Molecular Formula C31H33N2NaO6S2
Molecular Weight (g/mol) 616.72
MDL Number MFCD00060075
InChI Key UWGCNDBLFSEBDW-UHFFFAOYSA-M
Synonym Acid Brilliant Green J, Acid Green 16, Lissamine Green V, Pontacyl Green NV
PubChem CID 160685
IUPAC Name sodium 4-{[4-(diethylamino)phenyl][4-(diethyliminiumyl)cyclohexa-2,5-dien-1-ylidene]methyl}naphthalene-2,7-disulfonate
SMILES [Na+].CCN(CC)C1=CC=C(C=C1)C(=C1C=CC(C=C1)=[N+](CC)CC)C1=CC(=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color Purple-Red
Quantity 5 g
Formula Weight 616.72
Physical Form Crystalline Powder
Chemical Name or Material Erio Green B
EINECSNumber (5)-1738
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.