Learn More
TARGETMOL CHEMICALS INC ERTUGLIFLOZIN 100MG
Supplier:  TARGETMOL CHEMICALS INC T4999
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-04971729 (Ertugliflozin (PF-04971729)) is a potent and selective inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2). purity: 99%
Catalog No. 50-257-9842
missing translation for 'QuantityLeft'