Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Escitalopram oxalate, Tocris Bioscience™

Catalog No. p-4816698 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective serotonin reuptake inhibitor (SSRI)

Escitalopram oxalate is a selective 5-HT reuptake inhibitor (Ki values are 0.89, 8150, and 10500 nM for serotonin, noradrenalin, and dopamine transporters respectively). Escitalopram oxalate is an S-enantiomer and eutomer of citalopram (Cat. No. 1427).

Chemical Identifiers

CAS 219861-08-2
Molecular Formula C22H23FN2O5
Molecular Weight (g/mol) 414.433
InChI Key KTGRHKOEFSJQNS-BDQAORGHSA-N
Synonym escitalopram oxalate, lexapro, cipralex, s-citalopram oxalate, citalopram oxalate, unii-5u85dbw7lo, 5u85dbw7lo, s-+-citalopram oxalate, s-1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile oxalate
PubChem CID 146571
IUPAC Name (1S)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid
SMILES CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.C(=O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 414.43
Percent Purity 98%
Chemical Name or Material Escitalopram oxalate
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.