Learn More
Bevco Precision Manufacturing Co Doral Mid Height ESD/ISO 5 Cleanroom Blue Vinyl Chair; Articulating Seat & Back Tilt; Polished Aluminum Base w/Adjustable Footring
Supplier:  Bevco Precision Manufacturing Co 5351E2BLU
Encompass_Preferred
BEVCO’s Doral ESD/ISO 5 Cleanroom Series models are designed to meet specific particulate goals of ISO 14644-1 and the ESD requirements of ANSI/ESD STM12. 1-2019 delivering a combination ESD/Cleanroom seating solution for highly technical environments. This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations. Fully adjustable production-style back has a durable scuff resistant back pan to protect the upholstery. A waterfall seat promotes lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 19" - 26. 5". ESD blue vinyl meets CAL 117 standards. The chair features a sturdy 27" diameter polished aluminum base with adjustable ESD footring. Model comes standard with ESD mushroom glides. Optional ESD casters can be ordered separately.
Catalog No. NC2023035
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.