Learn More
Transforming Technologies™ ESD Jacket with Lapel Collar and Snap Cuff
SDP
Catalog No. p-200020838
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9010 Series ESD Jackets with Lapel collar and knit cuff provide high-quality ESD protection are are fabricated from lightweight 90% polyester and 10% carbon fabric for unmatched operator comfort. These jackets feature a snap-front closure and three roomy patch pockets.
  • Medium weight ESD jacket ideal for all working environments and numerous applications
  • Made with highly durable materials and maintains consistent continuity readings, even after 100 wash cycles
  • Certified a Level 3 ESD Jacket (A Groundable Garment System)
Specifications
Size X-Small
Gender Unisex
Material Polyester
Color Black
Closure Type Snap
Certifications/Compliance ANSI ESD S202.0
Collar Type Lapel
Cuff Style Snap
Disposable Reusable
Latex Free Yes
No. of Pockets 3
Resistance <1 x 106
Product Type ESD Jacket
Sleeve Style 3/4 Length
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.