Learn More
Ethanol, 200 proof (100%), USP, Decon™ Labs
Shop All Decon Labs Products

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Melting Point
-114°C
CAS Min %
99.9
CAS Max %
100.0
Density
6.62
Boiling Point
78.3°C
Quantity
0.125 gal.
Packaging
Glass Bottle
Physical Form
Liquid
UN Number
UN1170
Grade
BP/EP/FCC/JP/NF/USP
DOTInformation
DOT Class: 3, Packaging II
Recommended Storage
Do not store Ethanol at temperature above 120°F
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.