Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 99.5%, anhydrous, 200 proof, Thermo Scientific Chemicals
GSA_VA

Catalog No. AC615101000
Click to view available options
Quantity:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS

64-17-5, 7732-18-5

Molecular Formula

C2H6O

Molecular Weight (g/mol)

46.069

MDL Number

MFCD00003568

InChI Key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Synonym

ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol

PubChem CID

702

ChEBI

CHEBI:16236

IUPAC Name

ethanol

SMILES

CCO

Specifications

Melting Point

-114°C

Density

0.7900g/mL

Boiling Point

78°C

Quantity

100 mL

Flash Point

12°C (53.6°F)

Infrared Spectrum

Conforms to reference

Refractive Index

1.3585 to 1.3605

Assay Percent Range

99.5%

Residue after Evaporation

0.0025% max.

Specific Gravity

0.7964 Max.

Titratable Acid

0.0005 meq/g max.

Linear Formula

CH3CH2OH

Titratable Base

0.0002 meq/g max.

Water

0.05% max.

Merck Index

15, 3814

Formula Weight

46.07

Percent Purity

≥99.5%

Chemical Name or Material

Ethanol, Anhydrous, 200 proof

Hazard Category

Danger

Hazard Statement

GHS H Statement:
Highly flammable liquid and vapor.

Precautionary Statement

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.