Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ether, Preserved with about 2% Alcohol, 0.2% max Water and 7ppm min BHT, U.S.P., J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
250mL
1L
Packaging:
Aluminum Bottle
Specifications
Specifications
Flash Point | −45.00°C |
Packaging | Aluminum Bottle |
Quantity | 250mL |
Specific Gravity | 1L = 0.71kg |
Formula Weight | 74.12 |
Grade | USP |
Chemical Name or Material | Ether, Preserved with about 2% Alcohol, 0.2% max Water and 7ppm min BHT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction