Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Hexylacetoacetate 95.0+%, TCI America™
SDP

Catalog No. H012925ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 29214-60-6
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.305
MDL Number MFCD00026889
InChI Key BRGRZPQESQKATK-UHFFFAOYSA-N
Synonym 2-Acetyl-n-octanoic Acid Ethyl Ester, Ethyl 2-Acetyl-n-octanoate, Hexylacetoacetic Acid Ethyl Ester
PubChem CID 121790
IUPAC Name ethyl 2-acetyloctanoate
SMILES CCCCCCC(C(=O)C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 149°C
Quantity 25 mL
Formula Weight 214.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Hexylacetoacetate
EINECSNumber (2)-1505&(2)-2763
RTECSNumber RH0352500
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.