Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-chloro-2,2-dimethylpropanoate, ≥97%, Thermo Scientific™

Catalog No. RH00660EA
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

106315-37-1

Molecular Formula

C7H13ClO2

Molecular Weight (g/mol)

164.629

MDL Number

MFCD00220052

InChI Key

NEVHNBRNUWSFSB-UHFFFAOYSA-N

PubChem CID

287572

IUPAC Name

ethyl 3-chloro-2,2-dimethylpropanoate

SMILES

CCOC(=O)C(C)(C)CCl

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Quantity

10 g

Synonym

3-chloro-2,2-dimethyl-propionic acid ethyl ester, propanoic acid,3-chloro-2,2-dimethyl-, ethyl ester, acmc-1c8oo, ethyl 3-chloro-2,2-dimethyl-propanoate, propanoic acid, 3-chloro-2,2-dimethyl-, ethyl ester

Formula Weight

164.63

Percent Purity

97%

Chemical Name or Material

Ethyl 3-chloro-2, 2-dimethylpropanoate

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

Precautionary Statement

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.