Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-(4-fluorophenyl)-2-methyl-3-furoate, 95%, Thermo Scientific™

Catalog No. p-3075902
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 111787-83-8
Molecular Formula C14H13FO3
Molecular Weight (g/mol) 248.25
MDL Number MFCD05664426
InChI Key XUYCJNMLONEKAS-UHFFFAOYSA-N
Synonym ethyl 5-4-fluorophenyl-2-methyl-3-furoate, ethyl 5-4-fluorophenyl-2-methylfuran-3-carboxylate, 3-furancarboxylic acid,5-4-fluorophenyl-2-methyl-, ethyl ester, acmc-20mesw, 2-methyl-5-4-fluorophenyl furan-3-carboxylic acid ethyl ester
PubChem CID 2794727
IUPAC Name ethyl 5-(4-fluorophenyl)-2-methylfuran-3-carboxylate
SMILES CCOC(=O)C1=C(C)OC(=C1)C1=CC=C(F)C=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 248.25
Chemical Name or Material Ethyl 5-(4-fluorophenyl)-2-methyl-3-furoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.