Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate dihydrochloride 0.5 hydrate, 97%, Thermo Scientific™

Catalog No. p-3522575
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 623564-18-1
Molecular Formula C9H15Cl2N3O2
Molecular Weight (g/mol) 268.14
MDL Number MFCD17015229
InChI Key UVYMXIAFTVXRIT-UHFFFAOYSA-N
Synonym ethyl 5h,6h,7h,8h-imidazo 1,2-a pyrazine-2-carboxylate dihydrochloride, ethyl 5,6,7,8-tetrahydroimidazo 1,2-a pyrazine-2-carboxylate-hydrogen chloride 1/2, ethyl 5,6,7,8-tetrahydroimidazo 1,2-a pyrazine-2-carboxylate dihydrochloride 0.5 hydrate
PubChem CID 24229538
IUPAC Name ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate;dihydrochloride
SMILES Cl.Cl.CCOC(=O)C1=CN2CCNCC2=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 277.15
Percent Purity 97%
Chemical Name or Material Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate dihydrochloride 0.5 hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.