Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-amino-1-methylpyrazole-4-carboxylate, 97%, Thermo Scientific™

Catalog No. EN00291FA
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

31037-02-2

Molecular Formula

C7H11N3O2

Molecular Weight (g/mol)

169.18

MDL Number

MFCD00051652

InChI Key

MEUSJJFWVKBUFP-UHFFFAOYSA-N

Synonym

ethyl 5-amino-1-methyl-1h-pyrazole-4-carboxylate, 5-amino-1-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl5-amino-1-methylpyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid, 5-amino-1-methyl-, ethyl ester, 5-amino-1-methylpyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-1-methyl-4-pyrazolecarboxylate, ethyl 5-amino-1-methyl-pyrazole-4-carboxylate, acmc-1agpk, maybridge1_003833, ethyl-5-amino-1-methyl-1h-pyrazol-4-carboxylat

PubChem CID

271299

SMILES

CCOC(=O)C1=C(N)N(C)N=C1

Specifications

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Quantity

100 g

IUPAC Name

ethyl 5-amino-1-methylpyrazole-4-carboxylate

Formula Weight

169.18

Percent Purity

97%

Chemical Name or Material

Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.