Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ethyl Alcohol, 95% Denatured w/MIBK
SCI_ED

Catalog No. p-4573435 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
3.8 L
Packaging:
95% Denatured w/MIBK
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Quantity 500 mL
Packaging 95% Denatured w/MIBK
Physical Form Liquid
Linear Formula C2H5OH
Formula Weight 46.07
Chemical Name or Material Ethyl Alcohol
ShelfLife 5 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.