Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains 2% ethanol, >99.9% (excluding ethanol), Honeywell™ Burdick & Jackson

Catalog No. p-7111741 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4 x 4L
6 x 1 L
Packaging:
Glass Bottle

Chemical Identifiers

CAS 60-29-7, 64-17-5
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Absorbance 0.08 AU max. at 250 nm (UV), 0.01 AU max. at 275 nm (UV), 0.005 AU max. at 300 nm (UV)
CAS Max % 2.0000%, 98.0000%
UN Number UN1155
Beilstein 1696894
Formula Weight 74.12
Percent Purity >99.9%
Grade HPLC
Density 0.713 g/cm3
Packaging Glass Bottle
Flash Point −45°C
Boiling Point 35°C
Quantity 4 x 4L
Assay GC (excluding ethanol)
Chemical Name or Material Ethyl Ether, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains 2% ethanol, >99.9% (excluding ethanol)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.