Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Heptafluorobutyrate 97.0+%, TCI America™
SDP

Catalog No. p-7130627 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 356-27-4
Molecular Formula C6H5F7O2
Molecular Weight (g/mol) 242.093
MDL Number MFCD00000434
InChI Key JVHJRIQPDBCRRE-UHFFFAOYSA-N
Synonym ethyl heptafluorobutyrate, ethyl perfluorobutyrate, heptafluorobutyric acid ethyl ester, butanoic acid, heptafluoro-, ethyl ester, ethyl heptafluorobutanoate, ethyl perfluorobutanoate, ethyl heptafluoro-n-butyrate, butyric acid, heptafluoro-, ethyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, ethyl ester, pubchem12539
PubChem CID 9644
IUPAC Name ethyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES CCOC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 96°C
Quantity 5 g
UN Number 3272
Formula Weight 242.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Heptafluorobutyrate
RTECSNumber ET4200000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.