Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl p-Anisate 99.0+%, TCI America™
SDP

Catalog No. p-7117138 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
100 mL
500mL

Chemical Identifiers

CAS 94-30-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00015151
InChI Key FHUODBDRWMIBQP-UHFFFAOYSA-N
Synonym ethyl p-anisate, ethyl anisate, ethyl p-methoxybenzoate, p-anisic acid, ethyl ester, benzoic acid, 4-methoxy-, ethyl ester, 4-methoxybenzoic acid ethyl ester, benzoic acid, p-methoxy-, ethyl ester, ethyl p-anisoate, unii-kj95h2s7nm, p-anisic acid ethyl ester
PubChem CID 60979
IUPAC Name ethyl 4-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)OC

Specifications

Color Colorless
Boiling Point 143°C
Quantity 25 mL
Formula Weight 180.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl p-Anisate
EINECSNumber (3)-3054
RTECSNumber BZ4697000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.