Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle2-amino-4-methylthiophene-3-carboxylate, 97%, Thermo Scientific™

Catalog No. GK00116DA
Click to view available options
Quantity:

Chemical Identifiers

CAS

43088-42-2

Molecular Formula

C8H11NO2S

Molecular Weight (g/mol)

185.241

MDL Number

MFCD00051669

InChI Key

ILYCZKOBLRJJSW-UHFFFAOYSA-N

Synonym

2-amino-4-methylthiophene-3-carboxylic acid ethyl ester, 2-amino-4-methyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl2-amino-4-methylthiophene-3-carboxylate, ethyl 2-amino-4-methyl-3-thiophenecarboxylate, acmc-20a9ux, maybridge1_003755, 2-amino-4-methyl-3-ethoxycarbonyl-thiophene, ethyl 2-amino4-methylthiophene-3-carboxylate, 2-amino-3-ethoxycarbonyl-4-methylthiophene

PubChem CID

521132

IUPAC Name

ethyl 2-amino-4-methylthiophene-3-carboxylate

SMILES

CCOC(=O)C1=C(SC=C1C)N

Specifications

CAS Min %

96.5

CAS Max %

100.0

Assay Percent Range

97%

Quantity

1 g

Formula Weight

185.25

Percent Purity

97%

Chemical Name or Material

Ethyl 2-amino-4-methylthiophene-3-carboxylate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.