Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, 98.0%, MilliporeSigma™

Catalog No. 50947895 Shop All MilliporeSigma Products
Click to view available options
Quantity:
4 kg

Chemical Identifiers

CAS 107-21-1
Molecular Weight (g/mol) 62.07

Specifications

pH 6 to 7.5 (100 g/L, H2O, 20°C)
Melting Point −14 to −10°C
Boiling Point 197.6°C (1013 hPa)
Quantity 4 kg
Flash Point 111°C
Packaging Glass Bottle (Pre-packed)
Assay Percent Range 98% min
Vapor Pressure 0.053 hPa (20°C)
Solubility Information 1000 g/L
Recommended Storage Store at room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.