Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol, 99% min., MP Biomedicals™

Catalog No. p-4404786 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
250 mL
1 L

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Color Colorless
Density 1.113g/cm3 at 20°C
Quantity 250 mL
Physical Form Liquid
Assay Percent Range 99% min.
Formula Weight 62.07
Chemical Name or Material Ethylene glycol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.