Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Eticlopride hydrochloride

Catalog No. p-7112863 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

D2 and D3 antagonist (D3 > D2)

Eticlopride hydrochloride is a selective dopamine D2/D3 receptor antagonist (Ki values are 0.50 and 0.16 nM respectively). Antipsychotic.

Specifications

Chemical Name or Material 3-Chloro-5-ethyl-N-[[(2S)-1-ethyl-2-pyrrolidinyl)methyl]-6-hydroxy-2-methoxy-benzamide hydrochloride
CAS 97612-24-3
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in water
Molecular Formula C17H25ClN2O3.HCl
Formula Weight Observed MW: 377.31

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.