Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etilefrine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. p-7128971 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 943-17-9
Molecular Formula C10H16ClNO2
Molecular Weight (g/mol) 217.69
MDL Number MFCD00060167
InChI Key KTNROWWHOBZQGK-UHFFFAOYNA-N
Synonym 2-Ethylamino-1-(3-hydroxyphenyl)ethanol Hydrochloride
PubChem CID 164652
IUPAC Name hydrogen 3-[2-(ethylamino)-1-hydroxyethyl]phenol chloride
SMILES [H+].[Cl-].CCNCC(O)C1=CC=CC(O)=C1

Specifications

Melting Point 122°C
Color White
Quantity 5 g
Formula Weight 217.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Etilefrine Hydrochloride
EINECSNumber (3)-1060&(9)-0225
RTECSNumber DO5618000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.