Learn More
Electron Microscopy Sciences Evaporated Pt/Ir On Holey Carbon
Evaporated Pt/Ir on Holey carbon film. Holey carbon film support provides holes for ease of focus and astigmatism correction. The dots of evaporated Pt/Ir provide dense particles for resolution checks through the particle seperation test.