Learn More
TARGETMOL CHEMICALS INC EVODENOSON 5MG
Supplier:  TARGETMOL CHEMICALS INC T31731
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Evodenoson (ATL313) is a potent and selective adenosine A2a receptor (A2aR) agonist for the treatment of eye diseases tumors and immune system disorders and may be used in studies of open-angle glaucoma and blood tumors. purity: 99%
Catalog No. 50-258-0353
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.