Learn More
Thermo Scientific™ eVol™ XR Dispensing System Charger and Stand
Recharge your Thermo Scientific™ eVol™ Dispensing System with this charger, and get the stand that holds your eVol Dispensing System when not in use.
Supplier: Thermo Scientific™ 66002027
Description
Order the Thermo Scientific™ eVol™ Dispensing System Charger and Stand to recharge your dispensing system. The eVol dispenser is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:- Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
- Preparation of calibration standards
- Addition of internal standards
Repeatable Performance for Higher Confidence in Your Results
- Can be calibrated easily by a user to ensure validity of results
- Intuitive, easy-to-use interface with touch wheel and full-color screen
- Complies with GLP and GMP guidelines
- Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
- Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
Specifications
Charger for eVol system, 1 pk | |
Dispensing System Charger and Stand | |
HyperSep | |
Electronic Dispensing System |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.