Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EX 527, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
EX 527 is a selective inhibitor of SIRT1 that does not inhibit histone deacetylase (HDAC) or other sirtuin deacetylase family members (IC50 values are 98, 19600, 48700, > 100000 and > 100000 nM for SIRT1, SIRT2, SIRT3, HDAC and NADase respectively). In an animal model of Huntington's disease, EX 527 rescues neuronal degeneration, extends lifespan, reduces huntingtin inclusions and prevents ventricular enlargement. EX 527 enhances p53 acetylation in response to DNA damaging agents. Orally bioavailable and brain penetrant.
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 248.71 |
Percent Purity | >98% |
Chemical Name or Material | EX 527 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction