Learn More
Eurofins Discoverx PathHunter eXpress CXCR1 (IL8RA) U2OS β-Arrestin GPCR Assay, 2-Plate Kit. (93-0226E3CP0M)
Supplier: Eurofins Discoverx 930226E3CP0M
The PathHunter CXCR1 (IL8RA) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CXCL8/IL8) based activation of CXCR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR1 and EA-tagged β-Arrestin-2. Activation of CXCR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.