Learn More
Eurofins Discoverx PathHunter eXpress MC5R CHO-K1 β-Arrestin GPCR Assay, 2-Plate Kit. (93-0231E2CP2M)
Supplier:  Eurofins Discoverx 930231E2CP2M
Encompass
The PathHunter MC5R eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. α-MSH) based activation of MC5R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MC5R and EA-tagged β-Arrestin-2. Activation of MC5R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0013
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.