Learn More
Zymo Research Corporation EZ DNA Methylation™-Direct Kit, 50 Rxns.
Supplier: Zymo Research Corporation D5020
The EZ DNA Methylation™-Direct™ Kit is a further refinement of our popular EZ DNA Methylation™ and EZ DNA Methylation™-Gold™ kits. These products feature reliable and complete DNA bisulfite conversion directly from blood, tissue, and cells without the prerequisite for DNA purification. The increased sensitivity of these kits make it possible to amplify bisulfite-converted DNA from as few as 10 cells or 50 pg DNA. Like the EZ DNA Methylation™-Gold™ kits, DNA denaturation and bisulfite conversion processes are combined into a single step. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including restriction endonuclease digestion, sequencing, microarrays, etc.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.