Learn More
Air-Tite EZ FLO Hypodermic Needle
SDP
Smooth entry, smooth delivery
Supplier:  Air-Tite NEZ22100
 View more versions of this product
Catalog No. 14-817-231
missing translation for 'QuantityLeft'
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Air-Tite Product's newest addition, the EZ FLO line of hypodermic needles, offer high quality and a broad selection at an affordable price.
  • Ranging from 18 gauge to 30 gauge, these needles offer a standard bevel, thin wall from 18 g to 25 g and regular-wall in 27 g and 30 g to combine comfort and durability.
  • Each needle is sterile, individually packaged in a box of 100.
  • Made of surgical stainless steel and a polypropylene hub.
Specifications
Specifications
Surgical Stainless Steel
22 G
100/Box
Gray
1 in.
Sterile
Product Suggestions
Product Suggestions