Learn More
Alkali Scientific EZ-Punch™ Sealing Film, Sterile, 50/pk
Supplier: Alkali Scientific SF14MLS
EZ-Punch™ Sealing Film is designed for short-term processing and storage of samples that require multiple access to wells by pipet tips or robotic probes. A pre-cut “X” over each well creates bendable flaps that provide sample access without coring or adhesive fouling of tips or probes. Ideal for COVID-19 sample collection in deep-well plates using swabs. This pierceable film has printed circles that aid in the alignment of the film to the plate. Printed alphanumerics identify each well of a 96-well plate. Reseals after piercing for continued sample protection. To help position the film to the plate, align wells A-1 and H-12.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.