Learn More
Glass Expansion EzyLok 6mm Hose Adaptor
Supplier:  Glass Expansion 708080735
Encompass_Preferred
EzyLok 6mm Hose Adaptor. The EzyLok 6mm Hose Adaptor is an easy, safe, and effective way to connect soft walled argon supply tubes to your Glass Expansion F-Series and U-Series nebulizers when coupled with the EzyLok Connector for 4mm Tubing (QSM-4).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3017
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.