Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass. Bottom pad prevents chipping and breaking of pipette tips. All components can and will withstand repeated dry or steam heat serialization. Supplied stainless steel closure
SDP
Supplier:  Bellco Glass, Inc. 129038300
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom pad prevents chipping and breaking of pipette tips. All components can and will withstand repeated dry or steam heat serialization. Supplied stainless steel closure
Catalog No. 50-194-4672
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.