Learn More
Jackson Safety Face Shield Cap Adapters
Jackson Safety 282 B Cord Mounted Cap Adapters for Non-Slotted Caps features a polyethylene spark deflector. These cord-mounted, cap adapters, designed specifically for non-slotted caps are available with face shields (sold separately), that are secured with internal plastic lugs.
Supplier: Jackson Safety 14943
This item is not returnable.
View return policy
Description
- Model 282-B Cap Adaptor with Crown
- Vertical lock-in eliminates face shield creep
- Oversized crown for added safety
- Specially designed metal clips press over the hard hat rim for maximum stability and firm fit
- Features five easy-to-swivel lock mounting pins for universal windows
- An extremely popular cap adaptor
Specifications
Silver | |
Metal Clips | |
Universal |
Hard Hats | |
Hard Hat Cap Adapter with Crown |