Learn More
Jackson Safety Face Shield Cap Adapters
This adapter pivots up and down and secures the face shield in a variety of positions with integral plastic lugs. Designed to use with slotted caps, this adapter is ideal when the job requires a hard hat and face shield. Securely attaches to SC-6, Sentry III and Charger Hard Hats (sold separately).
Supplier: Jackson Safety 14944
Description
- Model 382 B Slotted Cap Adaptor with Crown, Dielectric or Model 437 Slotted Cap Adaptor, Dielectric
- Vertical lock-in eliminates face shield creep (Model 382)
- Oversized crown for added safety
- Made entirely of non-conductive materials (Dielectric)
- Attaches to SC-6, Sentry III, Charger, and many other slotted hard hat brands
- A popular style dielectric face shield hard hat adaptor bracket (Model 437)
- Curved injection molded channel fits over the hard hat bill>
Specifications
Black | |
Face Shield Hard Hat adapters | |
Slotted Cap Adapter with Crown | |
Regular Brim |
Slotted Hard Hats | |
Slotted | |
Universal |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.