Learn More
Revvity Health Sciences Inc Factor VIII (human) AlphaLISA Detection Kit, 5,000 assay points
Supplier:  Revvity Health Sciences Inc AL342F
Encompass
Formats: Our HV (100 assay point) kits allow you to run 100 wells in 96-well format, using a 100 ¿L reaction volume (10 ¿L of sample). Our 500 assay point kit allows you to run 500 wells in 96-well or 384-well format, using a 50 ¿L reaction volume (5 ¿L of sample). Our 5,000 assay point kit allows you to run 5,000 wells in 96-well or 384-well format, using a 50 ¿L reaction volume (5 ¿L of sample). Features: No-wash steps, no separation steps ELISA alternative technology Sensitive detection Broad sample compatibility Small sample volume Results in less than 3 hours Half the time of an ELISA assay Factor VIII is a blood clotting protein which is also called anti-hemophilic factor (AHF). Factor VIII is produced in the liver and in the endothelial cells of other organs and tissues. It circulates in the bloodstream as an inactive form complexed with von Willebrand factor (vWF). In damaged blood vessels Factor VIII disassociates from vWF to become active Factor VIII. Activated Factor VIII
Catalog No. 50-210-6056
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.