Learn More
Medchemexpress LLC HY-105117 5mg Medchemexpress, Fantofarone CAS:114432-13-2 Purity:>98%
Medchemexpress, HY-105117 5mg Fantofarone CAS:114432-13-2 Fantofarone is a highly potent Calcium Channel antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.