Learn More
NEOGEN Fastidious Anaerobe Agar, 500 g
This item has a minimum order quantity of 2 per supplier requirements
Fastidious Anaerobe Agar is used for the cultivation of anaerobic microorganisms and is not intended for use in the diagnosis of disease or other conditions in humans. A primary isolation medium capable of growing most clinically significant anaerobes. Developed by Lab M (Neogen Corporation), comparisons have shown this medium to be superior to other formulations as a primary isolation medium for fastidious organisms. The peptones included have been chosen for maximum growth stimulation. Starch and sodium bicarbonate act as de-toxification agents while hemin encourages pigment production in Porphyromonas melaninogenicus. Specific growth promoting agents are Cysteine for Fusobacterium necrophorum, Propionibacterium acne and Bacteriodes fragilis, arginine for Eubacterium spp. soluble pyrophosphate for Porph. gingivalis and Porph. asaccharolyticus.