Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ FastPack™ Inlet Kits

Includes all necessary parts for maintenance such as replacing the injection port (inlet) liner, the critical inlet seals, and the septum
Supplier: Restek™ 23258
Description
- Injection port maintenance should be performed on a routine basis, and prior to installing any capillary column
- FastPack Inlet Kit includes all necessary parts for maintenance such as replacing the injection port (inlet) liner, the critical inlet seals, and the septum
- For optimum column performance, the injection port (inlet) liner must be free of septum particles, sample residue, or ferrule fragments
- Peak shape degradation, poor reproducibility, sample decomposition, and ghost peaks all are associated with using a contaminated liner
- The critical seal must fit tightly around the inlet liner to prevent carrier gas from leaking around the outside of the liner
- Frequent septum replacement prevents fragmentation or leaks, and maintains a leak-tight seal to exclude air from the inlet
- Thermolite Plus septa are preconditioned for quicker instrument uptime
- Always use gloves and forceps when handling septa to avoid contamination
- The only way to maintain optimum performance is by frequently changing the inlet seal and ensuring the seal is leak tight.
Specifications
4 mm Topaz Single Taper Splitless with Wool | |
4 mm | |
Mylar Bag | |
5 Kits/Pk. |
11 mm | |
Multiple | |
Injection Port Maintenance | |
Agilent GCs |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction