Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fatostatin A, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of sterol regulatory element binding protein (SREBP); impairs the activation of SREBP-1 and SREBP-2. Exhibits antiproliferative effects in DU 145 cells independently of IGF-1 signaling (IC50 = 0.1 μM); reverses hyperglycemia in diabetic (ob/ob) mice. Cell permeable.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 375.33 |
Percent Purity | >97% |
Chemical Name or Material | Fatostatin A |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction