Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenbendazole, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. p-7157454 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum

F1359, 43210-67-9, C15H13N3O2S

Spectrum™ Chemical Fenbendazole, USP is used as a broad spectrum benzimidazole anthelmintic that guards against gastrointestinal parasites in animals, mostly cattle, sheep, dogs, cats and rabbits. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 43210-67-9
Molecular Formula C15H13N3O2S
Molecular Weight (g/mol) 299.35
InChI Key HDDSHPAODJUKPD-UHFFFAOYSA-N
IUPAC Name methyl N-[6-(phenylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=CC=C(SC3=CC=CC=C3)C=C2N1

Specifications

CAS Max % 100%
Ignition Residue 0.3%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 101%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.